missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-n-Butyl-N-phenylbenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60499.03

Code : NEW

Additional Details : CAS Number : 74805-35-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15446968

  • £358.49 / 1g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

N-n-Butyl-N-phenylbenzenesulfonamide
289.39
3149018
74805-35-9
MFCD00420919

Specifications

N-n-Butyl-N-phenylbenzenesulfonamide
1g
Liquid
289.39
3149018
Yellow
74805-35-9
97%
MFCD00420919
Videos
SDS