missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-n-Butyl-2-nitrobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60858.03

Code : NEW

Additional Details : CAS Number : 89840-64-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15477358

  • £358.49 / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

N-n-Butyl-2-nitrobenzenesulfonamide
258.29
3372015
89840-64-2
MFCD00577596

Specifications

N-n-Butyl-2-nitrobenzenesulfonamide
89840-64-2
97%
MFCD00577596
1g
Solid
258.29
3372015
Videos
SDS