missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-n-Butyl-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65795.03

Code : NEW

Additional Details : CAS Number : 4107-02-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15482629

  • £154.76 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

N-n-Butyl-2,2-diphenylacetamide
267.37
2122659
4107-02-2
MFCD00507372

Specifications

N-n-Butyl-2,2-diphenylacetamide
4107-02-2
95%
MFCD00507372
1g
Solid
267.37
2122659
Videos
SDS