missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(benzyloxycarbonyl)-1H-pyrazole-1-carboxamidine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7035871
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
152120-55-3
C20H18N4O4
378.388
MFCD02683516
NRBUVVTTYMTSKM-UHFFFAOYSA-N
n,n'-bis-z-1-guanylpyrazole, n,n'-bis benzyloxycarbonyl-1h-pyrazole-1-carboxamidine, n,n-bisbenzyloxycarbonyl-1-guanylpyrazole, benzyl benzyloxy carbonyl amino 1h-pyrazol-1-yl methylene carbamate, pyrazole z 2, pyrazol z 2, n,n'-bs-z-1-guanylpyrazole, n,n;-bis-z-1-guanylpyrazole, benzyl n-1z-benzyloxy carbonyl imino pyrazol-1-yl methyl carbamate, n,n'-di-cbz-1h-pyrazole-1-carbo
6371643
benzyl (NZ)-N-[phenylmethoxycarbonylamino(pyrazol-1-yl)methylidene]carbamate
C1=CC=C(C=C1)COC(=O)NC(=NC(=O)OCC2=CC=CC=C2)N3C=CC=N3
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
152120-55-3
C20H18N4O4
378.388
MFCD02683516
NRBUVVTTYMTSKM-UHFFFAOYSA-N
n,n'-bis-z-1-guanylpyrazole, n,n'-bis benzyloxycarbonyl-1h-pyrazole-1-carboxamidine, n,n-bisbenzyloxycarbonyl-1-guanylpyrazole, benzyl benzyloxy carbonyl amino 1h-pyrazol-1-yl methylene carbamate, pyrazole z 2, pyrazol z 2, n,n'-bs-z-1-guanylpyrazole, n,n;-bis-z-1-guanylpyrazole, benzyl n-1z-benzyloxy carbonyl imino pyrazol-1-yl methyl carbamate, n,n'-di-cbz-1h-pyrazole-1-carbo
6371643
benzyl (NZ)-N-[phenylmethoxycarbonylamino(pyrazol-1-yl)methylidene]carbamate
C1=CC=C(C=C1)COC(=O)NC(=NC(=O)OCC2=CC=CC=C2)N3C=CC=N3

N,N'-Bis(benzyloxycarbonyl)-1H-pyrazole-1-carboxamidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Bis(benzyloxycarbonyl)-1H-pyrazole-1-carboxamidine is used as pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 87°C to 91°C
Quantity 1 g
Formula Weight 378.38
Percent Purity ≥98%
Chemical Name or Material N,N'-Bis(benzyloxycarbonyl)-1H-pyrazole-1-carboxamidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'