missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Bis(acryloyl)cystamine, Electrophoresis Reagent, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
BAC is used as a reversible cross-linker for polyacrylamide gels, to simplify isolation of macromolecules separated on the gels. The disulfide linkage can be broken with a suitable reducing agent. A 1-g slice of a 4% (12:1) gel will dissolve in 5 mL of 100 mM DTT in TBE (pH 8.3) in less than 30 min. The pH of 8.3 is optimum for rapid dissolution of the gel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
BAC is used as a reversible cross-linker for polyacrylamide gels, to simplify isolation of macromolecules separated on the gels. The disulfide linkage can be broken with a suitable reducing agent. A 1-g slice of a 4% (12:1) gel will dissolve in 5 mL of 100 mM DTT in TBE (pH 8.3) in less than 30 min. The pH of 8.3 is optimum for rapid dissolution of the gel.
Solubility
Soluble at 50 mg/ml in acetic acid
Notes
Store at -20°C. Incompatible with heat, oxidizing agents.
BAC is used as a reversible cross-linker for polyacrylamide gels, to simplify isolation of macromolecules separated on the gels. The disulfide linkage can be broken with a suitable reducing agent. A 1-g slice of a 4% (12:1) gel will dissolve in 5 mL of 100 mM DTT in TBE (pH 8.3) in less than 30 min. The pH of 8.3 is optimum for rapid dissolution of the gel.
Solubility
Soluble at 50 mg/ml in acetic acid
Notes
Store at -20°C. Incompatible with heat, oxidizing agents.
Specifications
Melting Point | 123°C to 124°C |
Color | White |
Quantity | 25 g |
Solubility Information | Soluble at 50mg/mL in acetic acid |
Formula Weight | 260.37 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | N,N'-Bis(acryloyl)cystamine, Electrophoresis Reagent |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'