Learn More
N-MPPP Hydrochloride, Tocris Bioscience™
Selective κ agonist
Brand:  Tocris (Bio-Techne) 078350
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13428003
  • £352.80 / 50mg
Estimated Shipment date
from Supplier 25-03-2020
Description
Description
High affinity κ agonist with no measured binding at μ or δ sites.
Specifications
Chemical Identifiers
207452-97-9
358.91
n-mppp hydrochloride, n-methyl-n-1s-1-phenyl-2-1-pyrrolidinyl ethyl phenylacetamide hydrochloride
N-methyl-2-phenyl-N-(1-phenyl-2-pyrrolidin-1-ylethyl)acetamide;hydrochloride
C21H27ClN2O
BJJKMJWQLJIRGF-UHFFFAOYSA-N
53412424
CN(C(CN1CCCC1)C2=CC=CC=C2)C(=O)CC3=CC=CC=C3.Cl
Specifications
50mg
207452-97-9
n-mppp hydrochloride, n-methyl-n-1s-1-phenyl-2-1-pyrrolidinyl ethyl phenylacetamide hydrochloride
CN(C(CN1CCCC1)C2=CC=CC=C2)C(=O)CC3=CC=CC=C3.Cl
358.91
358.91
N-MPPP Hydrochloride
C21H27ClN2O
BJJKMJWQLJIRGF-UHFFFAOYSA-N
N-methyl-2-phenyl-N-(1-phenyl-2-pyrrolidin-1-ylethyl)acetamide;hydrochloride
53412424
>98%