missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpyrrole-2-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number 10522734
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1192-58-1
C6H7NO
109.13
MFCD00003087
OUKQTRFCDKSEPL-UHFFFAOYSA-N
1-methyl-1h-pyrrole-2-carbaldehyde, 1-methylpyrrole-2-carboxaldehyde, n-methylpyrrole-2-carboxaldehyde, n-methyl-2-pyrrolecarboxaldehyde, 1-methyl-1h-pyrrole-2-carboxaldehyde, 2-formyl-1-methylpyrrole, 1-methyl-2-pyrrolecarboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 1-methyl, unii-m0hyh3d7sx, 1-methyl-2-formylpyrrole
14504
1-methylpyrrole-2-carbaldehyde
CN1C=CC=C1C=O
This item is not returnable. View return policy
1192-58-1
C6H7NO
109.13
MFCD00003087
OUKQTRFCDKSEPL-UHFFFAOYSA-N
1-methyl-1h-pyrrole-2-carbaldehyde, 1-methylpyrrole-2-carboxaldehyde, n-methylpyrrole-2-carboxaldehyde, n-methyl-2-pyrrolecarboxaldehyde, 1-methyl-1h-pyrrole-2-carboxaldehyde, 2-formyl-1-methylpyrrole, 1-methyl-2-pyrrolecarboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 1-methyl, unii-m0hyh3d7sx, 1-methyl-2-formylpyrrole
14504
1-methylpyrrole-2-carbaldehyde
CN1C=CC=C1C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Density 1.0100g/mL
Boiling Point 87°C to 90°C (22.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5580 to 1.562
Quantity 100 g
Beilstein 21, I, 279
Specific Gravity 1.01
Formula Weight 109.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Methylpyrrole-2-carboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • N-Methylpyrrole-2-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'