missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpyrrole-2-carboxaldehyde, 98%, ACROS Organics™

Brand:  Acros Organics 156930250

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 1192-58-1 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10645592

  • £59.30 / 25g


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

N-Methylpyrrole-2-carboxaldehyde
97.5
≥97.5% (GC)
C6H7NO
MFCD00003087
1192-58-1
100.0
98%
109.13

Specifications

N-Methylpyrrole-2-carboxaldehyde
1192-58-1
100.0
Authentic
1.5580 to 1.5630 (20°C, 589nm)
≥97.5% (GC)
C6H7NO
MFCD00003087
Clear
25g
97.5
1.0100g/mL
Glass bottle
Liquid
98%
109.13
21, I, 279
Safety and Handling

Safety and Handling

Product Identifier
  • N-Methylpyrrole-2-carboxaldehyde

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documentation

Technical Documents