missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpropylamine, 96%, Thermo Scientific Chemicals

Catalog Number p-167786
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
627-35-0
C4H11N
73.14
MFCD00009361
GVWISOJSERXQBM-UHFFFAOYSA-N
n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm
12315
N-methylpropan-1-amine
CCCNC
This item is not returnable. View return policy
627-35-0
C4H11N
73.14
MFCD00009361
GVWISOJSERXQBM-UHFFFAOYSA-N
n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm
12315
N-methylpropan-1-amine
CCCNC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Color Colorless
Density 0.7100g/mL
Boiling Point 61.0°C to 63.0°C
Flash Point <−34°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2NHCH3
Refractive Index 1.3880 to 1.3910
Quantity 1 g
Specific Gravity 0.71
Formula Weight 73.14
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material N-Methylpropylamine, 96%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'