missing translation for 'onlineSavingsMsg'
Learn More

n-Methyloctylamine, 98%, Thermo Scientific Chemicals

Catalog Number 10771731
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2439-54-5
C9H21N
143.27
MFCD00048927
SEGJNMCIMOLEDM-UHFFFAOYSA-N
n-methyloctylamine, methyl octyl amine, n-methyl-n-octylamine, 1-octanamine, n-methyl, octylmethylamine, methyloctylamine, unii-4o0o17jz7d, n-methyl octylamine, n-methyl-octylamine, methyl n-octyl amine
75538
N-methyloctan-1-amine
CCCCCCCCNC
This item is not returnable. View return policy
2439-54-5
C9H21N
143.27
MFCD00048927
SEGJNMCIMOLEDM-UHFFFAOYSA-N
n-methyloctylamine, methyl octyl amine, n-methyl-n-octylamine, 1-octanamine, n-methyl, octylmethylamine, methyloctylamine, unii-4o0o17jz7d, n-methyl octylamine, n-methyl-octylamine, methyl n-octyl amine
75538
N-methyloctan-1-amine
CCCCCCCCNC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7900g/mL
Boiling Point 68.0°C (8.0 mmHg)
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7NHCH3
Refractive Index 1.4260 to 1.4280
Quantity 25 g
Specific Gravity 0.79
Formula Weight 143.27
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material n-Methyloctylamine
Show More Show Less
compliance-icons
Product Identifier
  • n-Methyloctylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'