Learn More
N-Methyllidocaine iodide, Tocris Bioscience™
Enhances biosynthesis of phosphatidylinositol
Brand:  Tocris (Bio-Techne) 104210
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13468363
  • £89.67 / 10mg
Estimated Shipment date
from Supplier 06-04-2020
Description
Description
Antiarrhythmic. Enhances the biosynthesis of phosphatidylinositol in hamster heart.
Specifications
Chemical Identifiers
1462-71-1
C15H25IN2O
n-methyllidocaine iodide, 2-2,6-dimethylphenyl amino-n,n-diethyl-n-methyl-2-oxoethanaminium iodide, n-methyllignocaine iodide, 2-2,6-dimethylanilino-2-oxoethyl-diethyl-methylazanium iodide, 2,6-dimethylphenyl carbamoyl methyl diethylmethylazanium iodide
Specifications
10mg
1462-71-1
n-methyllidocaine iodide, 2-2,6-dimethylphenyl amino-n,n-diethyl-n-methyl-2-oxoethanaminium iodide, n-methyllignocaine iodide, 2-2,6-dimethylanilino-2-oxoethyl-diethyl-methylazanium iodide, 2,6-dimethylphenyl carbamoyl methyl diethylmethylazanium iodide
>98%
N-Methyllidocaine iodide
C15H25IN2O
376.28