missing translation for 'onlineSavingsMsg'
Learn More

Nα-Methylhistamine dihydrochloride, Tocris Bioscience™

Non-selective H3 agonist

Brand:  Tocris (Bio-Techne) 057350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13457773

  • £329.28 / 50mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent histamine agonist, particularly at H3 receptors (potency relative to histamine is 81, 185, and 270% at H1, H2 and H3 respectively). Also displays agonist properties at H4 receptors to which it binds with moderate affinity (Ki = 23 nM).
Specifications

Chemical Identifiers

16503-22-3
198.091
nalpha-methylhistamine dihydrochloride, imidazole, 4-2-methylamino ethyl-, dihydrochloride, namh dihydrochloride, n-methyl-1h-imidazole-4-ethanamine dihydrochloride, n-methylhistamine dihydrochloride, n, a-methylhistamine dihydrochloride, n alpha-methylhistamine hcl, 2-1h-imidazol-4-yl ethyl methyl amine dihydrochloride, 2-3h-imidazol-4-yl ethyl methyl amine dihydrochloride, 1h-imidazole-4-ethanamine, n-methyl-, dihydrochloride
2-(1H-imidazol-5-yl)-N-methylethanamine;dihydrochloride
C6H13Cl2N3
AYUQICXJAMPXPF-UHFFFAOYSA-N
16078977
CNCCC1=CN=CN1.Cl.Cl

Specifications

50mg
16503-22-3
nalpha-methylhistamine dihydrochloride, imidazole, 4-2-methylamino ethyl-, dihydrochloride, namh dihydrochloride, n-methyl-1h-imidazole-4-ethanamine dihydrochloride, n-methylhistamine dihydrochloride, n, a-methylhistamine dihydrochloride, n alpha-methylhistamine hcl, 2-1h-imidazol-4-yl ethyl methyl amine dihydrochloride, 2-3h-imidazol-4-yl ethyl methyl amine dihydrochloride, 1h-imidazole-4-ethanamine, n-methyl-, dihydrochloride
CNCCC1=CN=CN1.Cl.Cl
198.091
198.09
Nα-Methylhistamine dihydrochloride
C6H13Cl2N3
AYUQICXJAMPXPF-UHFFFAOYSA-N
2-(1H-imidazol-5-yl)-N-methylethanamine;dihydrochloride
16078977
>98%
Videos
Documents