missing translation for 'onlineSavingsMsg'
Learn More

N-Methylhippuric acid, 99%, Thermo Scientific™

Catalog Number 10710561
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2568-34-5
C10H11NO3
193.20
MFCD00144940
PKCSYDDSNIJRIX-UHFFFAOYSA-N
benzoylsarcosine, n-methylhippuric acid, n-benzoyl-n-methylglycine, n-benzoylsarcosine, glycine, n-benzoyl-n-methyl, 2-n-methylbenzamido acetic acid, sarcosine, n-benzoyl, 2-benzoyl methyl amino acetic acid, chembl66391, 2-n-methyl-1-phenylformamido acetic acid
75728
2-[benzoyl(methyl)amino]acetic acid
CN(CC(O)=O)C(=O)C1=CC=CC=C1
This item is not returnable. View return policy
2568-34-5
C10H11NO3
193.20
MFCD00144940
PKCSYDDSNIJRIX-UHFFFAOYSA-N
benzoylsarcosine, n-methylhippuric acid, n-benzoyl-n-methylglycine, n-benzoylsarcosine, glycine, n-benzoyl-n-methyl, 2-n-methylbenzamido acetic acid, sarcosine, n-benzoyl, 2-benzoyl methyl amino acetic acid, chembl66391, 2-n-methyl-1-phenylformamido acetic acid
75728
2-[benzoyl(methyl)amino]acetic acid
CN(CC(O)=O)C(=O)C1=CC=CC=C1
CAS Min % 98.5
CAS Max % 100.0
Melting Point 101°C to 105°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5C(O)N(CH3)CH2COOH
Quantity 5 g
Formula Weight 193.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Methylhippuric acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • N-Methylhippuric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'