missing translation for 'onlineSavingsMsg'
Learn More

N-Methylformanilide, 99%, Thermo Scientific Chemicals

Catalog Number 10695362
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
10 kg
Packaging:
Glass Bottle
Plastic drum
This item is not returnable. View return policy
93-61-8
MFCD00003283
JIKUXBYRTXDNIY-UHFFFAOYSA-N
n-methylformanilide, formamide, n-methyl-n-phenyl, n-formyl-n-methylaniline, formanilide, n-methyl, methylphenylformamide, n-phenyl-n-methylformamide, n-methyl-n-formylaniline, n-methylformanilid, methyl phenyl formamide, n-methyl-n-phenyl-formamide
66737
N-methyl-N-phenylformamide
CN(C=O)C1=CC=CC=C1
This item is not returnable. View return policy
93-61-8
MFCD00003283
JIKUXBYRTXDNIY-UHFFFAOYSA-N
n-methylformanilide, formamide, n-methyl-n-phenyl, n-formyl-n-methylaniline, formanilide, n-methyl, methylphenylformamide, n-phenyl-n-methylformamide, n-methyl-n-formylaniline, n-methylformanilid, methyl phenyl formamide, n-methyl-n-phenyl-formamide
66737
N-methyl-N-phenylformamide
CN(C=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 8.0°C to 13.0°C
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 243.0°C to 244.0°C
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Molecular Formula C8H9NO
Linear Formula HCON(CH3)C6H5
Refractive Index 1.5600 to 1.5620
Quantity 5 g
Specific Gravity 1.09
Solubility Information Solubility in water: immiscible.
Molecular Weight (g/mol) 135.166
Formula Weight 135.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Methylformanilide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • N-Methylformanilide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'