missing translation for 'onlineSavingsMsg'
Learn More

N-Methyldiphenylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-162339
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
552-82-9
C13H13N
183.25
MFCD00041900
DYFFAVRFJWYYQO-UHFFFAOYSA-N
n-methyldiphenylamine, benzenamine, n-methyl-n-phenyl, methyldiphenylamine, n,n-diphenylmethylamine, diphenylamine, n-methyl, unii-b28zgh99ih, diphenyl methylamine, n-methyl-n-phenyl-aniline, b28zgh99ih, methyldiphenylamin
11098
N-methyl-N-phenylaniline
CN(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
552-82-9
C13H13N
183.25
MFCD00041900
DYFFAVRFJWYYQO-UHFFFAOYSA-N
n-methyldiphenylamine, benzenamine, n-methyl-n-phenyl, methyldiphenylamine, n,n-diphenylmethylamine, diphenylamine, n-methyl, unii-b28zgh99ih, diphenyl methylamine, n-methyl-n-phenyl-aniline, b28zgh99ih, methyldiphenylamin
11098
N-methyl-N-phenylaniline
CN(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -7.6°C
Color White to Beige
Density 1.0500g/mL
Boiling Point 296°C to 297°C
Flash Point >093°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5N(CH3)C6H5
Refractive Index 1.6215 to 1.6235
Quantity 5 g
Merck Index 15, 6126
Specific Gravity 1.05
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol
Formula Weight 183.25
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N-Methyldiphenylamine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • N-Methyldiphenylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'