missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-tert-butylamine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Density | 0.7270g/mL |
Boiling Point | 67.0°C to 69.0°C |
Flash Point | −26°C |
Assay Percent Range | 96% (GC) |
Packaging | Glass Bottle |
Linear Formula | (CH3)3CNHCH3 |
Refractive Index | 1.3950 to 1.3970 |
Quantity | 25 mL |
Show More |
Safety and Handling
Product Identifier
- N-Methyl-tert-butylamine
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'