missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-propargylbenzylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-3758465
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
555-57-7
C11H13N
159.23
MFCD00008576
DPWPWRLQFGFJFI-UHFFFAOYSA-N
pargyline, pargylamine, n-methyl-n-propargylbenzylamine, paragyline, eudatin, supirdyl, eutonyl, pargylin, n-methyl-n-2-propynylbenzylamine, benzyl-methyl-2-propinylamin
4688
CHEBI:7930
N-benzyl-N-methylprop-2-yn-1-amine
CN(CC#C)CC1=CC=CC=C1
This item is not returnable. View return policy
555-57-7
C11H13N
159.23
MFCD00008576
DPWPWRLQFGFJFI-UHFFFAOYSA-N
pargyline, pargylamine, n-methyl-n-propargylbenzylamine, paragyline, eudatin, supirdyl, eutonyl, pargylin, n-methyl-n-2-propynylbenzylamine, benzyl-methyl-2-propinylamin
4688
CHEBI:7930
N-benzyl-N-methylprop-2-yn-1-amine
CN(CC#C)CC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 0.9400g/mL
Boiling Point 86°C to 88°C (4.0 mmHg)
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5CH2N(CH3)CH2C∽CH
Refractive Index 1.5213
Quantity 1 g
Beilstein 12, II, 548
Merck Index 15, 7142
Specific Gravity 0.94
Solubility Information Solubility in water: soluble. Other solubilities: miscibel in ethanol,acetone
Formula Weight 159.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Methyl-N-propargylbenzylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Methyl-N-propargylbenzylamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'