missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Methyl-N-phenylbenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60784.03

Code : NEW

Additional Details : CAS Number : 90-10-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15407288

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Methyl-N-phenylbenzenesulfonamide
90-10-8
247.31
MFCD00514036

Specifications

N-Methyl-N-phenylbenzenesulfonamide
90-10-8
97%
MFCD00514036
1g
Solid
247.31
Videos
SDS