missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-n-octylamine, 97+%, Thermo Scientific Chemicals

Catalog Number p-7047306
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2439-54-5
C9H21N
143.274
MFCD00048927
SEGJNMCIMOLEDM-UHFFFAOYSA-N
n-methyloctylamine, methyl octyl amine, n-methyl-n-octylamine, 1-octanamine, n-methyl, octylmethylamine, methyloctylamine, unii-4o0o17jz7d, n-methyl octylamine, n-methyl-octylamine, methyl n-octyl amine
75538
N-methyloctan-1-amine
CCCCCCCCNC
This item is not returnable. View return policy
2439-54-5
C9H21N
143.274
MFCD00048927
SEGJNMCIMOLEDM-UHFFFAOYSA-N
n-methyloctylamine, methyl octyl amine, n-methyl-n-octylamine, 1-octanamine, n-methyl, octylmethylamine, methyloctylamine, unii-4o0o17jz7d, n-methyl octylamine, n-methyl-octylamine, methyl n-octyl amine
75538
N-methyloctan-1-amine
CCCCCCCCNC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.79
Boiling Point 68°C (8 mmHg)
Flash Point 63°C (145°F)
Linear Formula CH3(CH2)7NHCH3
Refractive Index 1.427
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 143.27
Percent Purity 98%
Chemical Name or Material N-Methyl-n-octylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'