missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-N-[(5-methyl-2-phenyl-3-furyl)methyl]amine, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Quantity | 250 mg |
Formula Weight | 201.27 |
Percent Purity | 90% |
Chemical Name or Material | N-methyl-N-[(5-methyl-2-phenyl-3-furyl)methyl]amine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'