missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-N-[(4-methyl-4H-thieno[3,2-b]pyrrol-5-yl)methyl]amine hydrochloride, 95%, Thermo Scientific™
Click to view available options
Amber Glass Bottle; 1g
Specifications
CAS | 335033-68-6 |
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Molecular Formula | C9H13ClN2S |
MDL Number | MFCD07772840 |
Quantity | 1 g |
Synonym | n-methyl-n-4-methyl-4h-thieno 3,2-b pyrrol-5-yl methyl amine hydrochloride, methyl 4-methylthieno 3,2-b pyrrol-5-yl methyl amine hydrochloride, methyl 4-methylthiopheno 2,3-d pyrrol-5-yl methyl amine, chloride, n-methyl-n-4-methyl-4h-theno 3,2-b pyrrol-5-yl methyl amne hydrochlorde, n-methyl-1-4-methyl-4h-thieno 3,2-b pyrrol-5-yl methanamine-hydrogen chloride 1/1 |
InChI Key | JUWOUDJRMHOUBO-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'