missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(3-pyridylmethyl)amine, 97%, Thermo Scientific™

Catalog Number p-2705706
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
20173-04-0
C7H10N2
122.171
MFCD00023610
MCSAQVGDZLPTBS-UHFFFAOYSA-N
n-methyl-1-pyridin-3-yl methanamine, n-methyl-n-3-pyridylmethyl amine, 3-methylaminomethyl pyridine, n-methyl pyridin-3-yl methanamine, 3-methylamino methyl pyridine, 3-pyridinemethanamine, n-methyl, 3-picolylmethylamine, n-methyl-3-pyridylmethylamine, methyl pyridin-3-ylmethyl amine, n-methyl-3-pyridinemethylamine
88393
N-methyl-1-pyridin-3-ylmethanamine
CNCC1=CN=CC=C1
This item is not returnable. View return policy
20173-04-0
C7H10N2
122.171
MFCD00023610
MCSAQVGDZLPTBS-UHFFFAOYSA-N
n-methyl-1-pyridin-3-yl methanamine, n-methyl-n-3-pyridylmethyl amine, 3-methylaminomethyl pyridine, n-methyl pyridin-3-yl methanamine, 3-methylamino methyl pyridine, 3-pyridinemethanamine, n-methyl, 3-picolylmethylamine, n-methyl-3-pyridylmethylamine, methyl pyridin-3-ylmethyl amine, n-methyl-3-pyridinemethylamine
88393
N-methyl-1-pyridin-3-ylmethanamine
CNCC1=CN=CC=C1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material N-methyl-N-(3-pyridylmethyl)amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'