missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-N-(3-pyridin-4-ylbenzyl)amine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 198.27 |
Percent Purity | 95% |
Chemical Name or Material | N-Methyl-N-(3-pyridin-4-ylbenzyl)amine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'