missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[3-(pyridin-2-yloxy)benzyl]amine, 97%, Thermo Scientific™

Catalog Number p-3522607
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
871825-60-4
C13H14N2O
214.27
MFCD09025856
OEPWSCXLHMBFPC-UHFFFAOYSA-N
n-methyl-n-3-pyridin-2-yloxy benzyl amine, methyl 3-pyridin-2-yloxy phenyl methyl amine, methyl 3-2-pyridyloxy phenyl methyl amine, benzenemethanamine,n-methyl-3-2-pyridinyloxy, n-methyl-1-3-pyridin-2-yloxy phenyl methanamine, n-methyl-1-3-pyridin-2-yl oxy phenyl methanamine
18525840
N-methyl-1-(3-pyridin-2-yloxyphenyl)methanamine
CNCC1=CC(OC2=CC=CC=N2)=CC=C1
This item is not returnable. View return policy
871825-60-4
C13H14N2O
214.27
MFCD09025856
OEPWSCXLHMBFPC-UHFFFAOYSA-N
n-methyl-n-3-pyridin-2-yloxy benzyl amine, methyl 3-pyridin-2-yloxy phenyl methyl amine, methyl 3-2-pyridyloxy phenyl methyl amine, benzenemethanamine,n-methyl-3-2-pyridinyloxy, n-methyl-1-3-pyridin-2-yloxy phenyl methanamine, n-methyl-1-3-pyridin-2-yl oxy phenyl methanamine
18525840
N-methyl-1-(3-pyridin-2-yloxyphenyl)methanamine
CNCC1=CC(OC2=CC=CC=N2)=CC=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 214.27
Percent Purity 97%
Chemical Name or Material N-methyl-N-[3-(pyridin-2-yloxy)benzyl]amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'