missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(2-thien-2-ylbenzyl)amine, 97%, Thermo Scientific™

Catalog Number p-3111069
Click to view available options
Quantity:
250 mg
5 g
This item is not returnable. View return policy
852180-66-6
C12H13NS
203.303
MFCD07368538
MRKJJEJYTBOUTH-UHFFFAOYSA-N
n-methyl-n-2-thien-2-ylbenzyl amine, n-methyl-1-2-thiophen-2-ylphenyl methanamine, n-methyl-2-thien-2-ylbenzylamine, methyl 2-thiophen-2-yl phenyl methyl amine, n-methyl-1-2-thiophen-2-yl phenyl methanamine, d0a2yg, n-methyl-n-2-thiophen-2-ylbenzyl amine, methyl 2-2-thienyl phenyl methyl amine, benzenemethanamine,n-methyl-2-2-thienyl, benzenemethanamine, n-methyl-2-2-thienyl
7060565
N-methyl-1-(2-thiophen-2-ylphenyl)methanamine
CNCC1=CC=CC=C1C2=CC=CS2
This item is not returnable. View return policy
852180-66-6
C12H13NS
203.303
MFCD07368538
MRKJJEJYTBOUTH-UHFFFAOYSA-N
n-methyl-n-2-thien-2-ylbenzyl amine, n-methyl-1-2-thiophen-2-ylphenyl methanamine, n-methyl-2-thien-2-ylbenzylamine, methyl 2-thiophen-2-yl phenyl methyl amine, n-methyl-1-2-thiophen-2-yl phenyl methanamine, d0a2yg, n-methyl-n-2-thiophen-2-ylbenzyl amine, methyl 2-2-thienyl phenyl methyl amine, benzenemethanamine,n-methyl-2-2-thienyl, benzenemethanamine, n-methyl-2-2-thienyl
7060565
N-methyl-1-(2-thiophen-2-ylphenyl)methanamine
CNCC1=CC=CC=C1C2=CC=CS2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 203.31
Percent Purity 95%
Chemical Name or Material N-methyl-N-(2-thien-2-ylbenzyl)amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'