missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[(1-methyl-1H-benzimidazol-2-yl)methyl]amine, 97%, Maybridge

Brand:  Maybridge CC03846DA

Code : 31

Additional Details : CAS Number : 137898-62-5 Weight : 0.05100kg Transport : UN number : 2923 Chem class : 8 Pack group : III

Packaging Amber Glass Bottle
Quantity 1g

 View more versions of this product

Product Code. 10401124

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

N-Methyl-N-[(1-methyl-1H-benzimidazol-2-yl)methyl]amine
96.5
97%
C10H13N3
MFCD08060627
137898-62-5
100.0
97%
175.23

Specifications

N-Methyl-N-[(1-methyl-1H-benzimidazol-2-yl)methyl]amine
96.5
Amber Glass Bottle
97%
175.23
1g
137898-62-5
100.0
97%
C10H13N3
MFCD08060627
Videos
SDS