missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Methyl-L-leucine benzyl ester p-toluenesulfonate salt, 95%

Brand:  Alfa Aesar™ H63349.06

Code : NEW

Additional Details : CAS Number : 42807-66-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15408266

  • £258.64 / 5g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

N-Methyl-L-leucine benzyl ester p-toluenesulfonate salt
407.52
N-Me-leu-OBzl p-Tos-OH
42807-66-9
MFCD00083659

Specifications

N-Methyl-L-leucine benzyl ester p-toluenesulfonate salt
Solid
407.52
N-Me-leu-OBzl p-Tos-OH
42807-66-9
95%
MFCD00083659
5g
Videos
SDS