missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(chroman-8-ylmethyl)amine, 97%, Thermo Scientific™

Catalog Number p-3881560
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
1048970-16-6
C11H15NO
177.247
MFCD12198126
VVCGFZWBAXPSIO-UHFFFAOYSA-N
n-methyl-chroman-8-ylmethyl amine, 3,4-dihydro-2h-1-benzopyran-8-ylmethyl methyl amine, chroman-8-ylmethyl methylamine, 1-3,4-dihydro-2h-1-benzopyran-8-yl-n-methylmethanamine
43811059
1-(3,4-dihydro-2H-chromen-8-yl)-N-methylmethanamine
CNCC1=CC=CC2=C1OCCC2
This item is not returnable. View return policy
1048970-16-6
C11H15NO
177.247
MFCD12198126
VVCGFZWBAXPSIO-UHFFFAOYSA-N
n-methyl-chroman-8-ylmethyl amine, 3,4-dihydro-2h-1-benzopyran-8-ylmethyl methyl amine, chroman-8-ylmethyl methylamine, 1-3,4-dihydro-2h-1-benzopyran-8-yl-n-methylmethanamine
43811059
1-(3,4-dihydro-2H-chromen-8-yl)-N-methylmethanamine
CNCC1=CC=CC2=C1OCCC2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 177.25
Percent Purity 97%
Chemical Name or Material N-methyl-(chroman-8-ylmethyl)amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'