missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(7-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)methylamine, 97%, Thermo Scientific™

Catalog Number p-3486376
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
886851-54-3
C8H10BrNO2S
264.137
MFCD08690308
FJHBRYOEMIQSPC-UHFFFAOYSA-N
n-methyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl methylamine, n-methyl-5-aminomethyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine, 7-bromo-2h,3h-thieno 3,4-b 1,4 dioxin-5-yl methyl methyl amine, 7-bromo 2h,3h-thiopheno 4,3-e 1,4-dioxan-5-yl methyl methylamine, thieno 3,4-b-1,4-dioxin-5-methanamine,7-bromo-2,3-dihydro-n-methyl, 1-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl-n-methylmethanamine
18525892
1-(5-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-7-yl)-N-methylmethanamine
CNCC1=C2C(=C(S1)Br)OCCO2
This item is not returnable. View return policy
886851-54-3
C8H10BrNO2S
264.137
MFCD08690308
FJHBRYOEMIQSPC-UHFFFAOYSA-N
n-methyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl methylamine, n-methyl-5-aminomethyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine, 7-bromo-2h,3h-thieno 3,4-b 1,4 dioxin-5-yl methyl methyl amine, 7-bromo 2h,3h-thiopheno 4,3-e 1,4-dioxan-5-yl methyl methylamine, thieno 3,4-b-1,4-dioxin-5-methanamine,7-bromo-2,3-dihydro-n-methyl, 1-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl-n-methylmethanamine
18525892
1-(5-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-7-yl)-N-methylmethanamine
CNCC1=C2C(=C(S1)Br)OCCO2
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 264.14
Percent Purity 90%
Chemical Name or Material N-methyl-(7-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'