missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-[6-(tetrahydropyran-4-yloxy)pyrid-3-yl]methylamine, 97%, Thermo Scientific™

Catalog Number p-3586872
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
910036-99-6
C12H18N2O2
222.288
MFCD09879908
CWRQCHDTRDIGCU-UHFFFAOYSA-N
n-methyl-6-tetrahydropyran-4-yloxy pyrid-3-yl methylamine, 5-methylamino methyl-2-tetrahydro-2h-pyran-4-yloxy pyridine, methyl 6-oxan-4-yloxy pyridin-3-yl methyl amine, n-methyl-1-6-oxan-4-yl oxy pyridin-3-yl methanamine, methyl 6-2h-3,4,5,6-tetrahydropyran-4-yloxy 3-pyridyl methyl amine, n-methyl-1-6-tetrahydro-2h-pyran-4-yl oxy pyridin-3-yl methanamine
24229584
N-methyl-1-[6-(oxan-4-yloxy)pyridin-3-yl]methanamine
CNCC1=CN=C(C=C1)OC2CCOCC2
This item is not returnable. View return policy
910036-99-6
C12H18N2O2
222.288
MFCD09879908
CWRQCHDTRDIGCU-UHFFFAOYSA-N
n-methyl-6-tetrahydropyran-4-yloxy pyrid-3-yl methylamine, 5-methylamino methyl-2-tetrahydro-2h-pyran-4-yloxy pyridine, methyl 6-oxan-4-yloxy pyridin-3-yl methyl amine, n-methyl-1-6-oxan-4-yl oxy pyridin-3-yl methanamine, methyl 6-2h-3,4,5,6-tetrahydropyran-4-yloxy 3-pyridyl methyl amine, n-methyl-1-6-tetrahydro-2h-pyran-4-yl oxy pyridin-3-yl methanamine
24229584
N-methyl-1-[6-(oxan-4-yloxy)pyridin-3-yl]methanamine
CNCC1=CN=C(C=C1)OC2CCOCC2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 222.29
Percent Purity 97%
Chemical Name or Material N-Methyl-[6-(tetrahydropyran-4-yloxy)pyrid-3-yl]methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'