missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-methyl-(5-pyrid-4-ylthien-2-yl)methylamine, 97%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Especificaciones
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 204.3 |
Percent Purity | 97% |
Chemical Name or Material | N-methyl-(5-pyrid-4-ylthien-2-yl)methylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'