missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(4-methyl-2-phenyl-1,3-oxazol-5-ylmethyl)amine, 97%, Maybridge™

£81.00 - £201.00

Chemical Identifiers

CAS 1031843-28-3
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.257
MDL Number MFCD11109322
InChI Key XCBYKJIAOCVNGY-UHFFFAOYSA-N
Synonym n-methyl-4-methyl-2-phenyl-1,3-oxazol-5-ylmethyl amine, methyl 4-methyl-2-phenyl-1,3-oxazol-5-yl methyl amine, 5-methylamino methyl-4-methyl-2-phenyl-1,3-oxazole, methyl 4-methyl-2-phenyl 1,3-oxazol-5-yl methyl amine, n-methyl-1-4-methyl-2-phenyl-1,3-oxazol-5-yl methanamine, n-methyl-1-4-methyl-2-phenyl-1,3-oxazol-5-yl methylamine
PubChem CID 33589464
IUPAC Name N-methyl-1-(4-methyl-2-phenyl-1,3-oxazol-5-yl)methanamine
SMILES CC1=C(OC(=N1)C2=CC=CC=C2)CNC
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10744755
SDS View Documents
Maybridge
CC18146CB
250mg Amber Glass Bottle
£81.00
250mg
View alternative products
Save Up to 
»
Estimated Shipment: 12-10-2021
Please to purchase this item. Need a web account? Register with us today!
10491464
SDS View Documents
Maybridge
CC18146DA
1g Amber Glass Bottle
£201.00
1g
View alternative products
Save Up to 
»
Estimated Shipment: 12-10-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

1031843-28-3
202.257
XCBYKJIAOCVNGY-UHFFFAOYSA-N
33589464
CC1=C(OC(=N1)C2=CC=CC=C2)CNC
C12H14N2O
MFCD11109322
n-methyl-4-methyl-2-phenyl-1,3-oxazol-5-ylmethyl amine, methyl 4-methyl-2-phenyl-1,3-oxazol-5-yl methyl amine, 5-methylamino methyl-4-methyl-2-phenyl-1,3-oxazole, methyl 4-methyl-2-phenyl 1,3-oxazol-5-yl methyl amine, n-methyl-1-4-methyl-2-phenyl-1,3-oxazol-5-yl methanamine, n-methyl-1-4-methyl-2-phenyl-1,3-oxazol-5-yl methylamine
N-methyl-1-(4-methyl-2-phenyl-1,3-oxazol-5-yl)methanamine

Specifications

N-Methyl-(4-methyl-2-phenyl-1, 3-oxazol-5-ylmethyl)amine
250mg
96.5
97%
MFCD11109322
XCBYKJIAOCVNGY-UHFFFAOYSA-N
N-methyl-1-(4-methyl-2-phenyl-1,3-oxazol-5-yl)methanamine
33589464
97%
Amber Glass Bottle
1031843-28-3
100.0
C12H14N2O
n-methyl-4-methyl-2-phenyl-1,3-oxazol-5-ylmethyl amine, methyl 4-methyl-2-phenyl-1,3-oxazol-5-yl methyl amine, 5-methylamino methyl-4-methyl-2-phenyl-1,3-oxazole, methyl 4-methyl-2-phenyl 1,3-oxazol-5-yl methyl amine, n-methyl-1-4-methyl-2-phenyl-1,3-oxazol-5-yl methanamine, n-methyl-1-4-methyl-2-phenyl-1,3-oxazol-5-yl methylamine
CC1=C(OC(=N1)C2=CC=CC=C2)CNC
202.257
202.26
SDS
Product Certifications