missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-4-(5-methyl-1,3,4-oxadiazol-2-yl)benzylamine, 95%, Maybridge

£81.00 - £111.00

Chemical Identifiers

CAS 944450-83-3
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.245
MDL Number MFCD11109326
InChI Key PBJZWGQLXKZEJT-UHFFFAOYSA-N
Synonym n-methyl-4-5-methyl-1,3,4-oxadiazol-2-yl benzylamine, methyl 4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methyl amine, 2-4-methyl aminomethyl phenyl-5-methyl-1,3,4-oxadiazole, methyl 4-5-methyl 1,3,4-oxadiazol-2-yl phenyl methyl amine, n-methyl-1-4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methanamine, n-methyl-1-4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methylamine
PubChem CID 26343642
IUPAC Name N-methyl-1-[4-(5-methyl-1,3,4-oxadiazol-2-yl)phenyl]methanamine
SMILES CC1=NN=C(O1)C2=CC=C(C=C2)CNC
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10377204 SDS View Documents Maybridge
CC61746CB
250mg Amber Glass Bottle £81.00
250mg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 03-06-2020
Please to purchase this item. Need a web account? Register with us today!
10143474 SDS View Documents Maybridge
CC61746DA
1g Amber Glass Bottle £111.00
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 03-07-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

944450-83-3
203.245
PBJZWGQLXKZEJT-UHFFFAOYSA-N
26343642
CC1=NN=C(O1)C2=CC=C(C=C2)CNC
C11H13N3O
MFCD11109326
n-methyl-4-5-methyl-1,3,4-oxadiazol-2-yl benzylamine, methyl 4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methyl amine, 2-4-methyl aminomethyl phenyl-5-methyl-1,3,4-oxadiazole, methyl 4-5-methyl 1,3,4-oxadiazol-2-yl phenyl methyl amine, n-methyl-1-4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methanamine, n-methyl-1-4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methylamine
N-methyl-1-[4-(5-methyl-1,3,4-oxadiazol-2-yl)phenyl]methanamine

Specifications

N-methyl-4-(5-methyl-1, 3, 4-oxadiazol-2-yl)benzylamine
250mg
94.5
95%
MFCD11109326
PBJZWGQLXKZEJT-UHFFFAOYSA-N
N-methyl-1-[4-(5-methyl-1,3,4-oxadiazol-2-yl)phenyl]methanamine
26343642
95%
Amber Glass Bottle
944450-83-3
100.0
C11H13N3O
n-methyl-4-5-methyl-1,3,4-oxadiazol-2-yl benzylamine, methyl 4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methyl amine, 2-4-methyl aminomethyl phenyl-5-methyl-1,3,4-oxadiazole, methyl 4-5-methyl 1,3,4-oxadiazol-2-yl phenyl methyl amine, n-methyl-1-4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methanamine, n-methyl-1-4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methylamine
CC1=NN=C(O1)C2=CC=C(C=C2)CNC
203.245
203.24