missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-[4-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methylamine, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Quantity | 1 g |
Formula Weight | 203.24 |
Percent Purity | 95% |
Chemical Name or Material | N-methyl-[4-(3-methyl-1, 2, 4-oxadiazol-5-yl)phenyl]methylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'