missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-4-(1-methyl-1H-pyrazol-3-yl)benzylamine, 90%, Thermo Scientific™

Catalog Number p-3541098
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
179873-47-3
C12H15N3
201.273
MFCD09702353
BBOCGPKUAXCMLG-UHFFFAOYSA-N
n-methyl-4-1-methyl-1h-pyrazol-3-yl benzylamine, benzenemethanamine,n-methyl-4-1-methyl-1h-pyrazol-3-yl, methyl 4-1-methylpyrazol-3-yl phenyl methyl amine, n-methyl-1-4-1-methyl-1h-pyrazol-3-yl phenyl methanamine, methyl 4-1-methyl-1h-pyrazol-3-yl phenyl methyl amine
24229474
N-methyl-1-[4-(1-methylpyrazol-3-yl)phenyl]methanamine
CNCC1=CC=C(C=C1)C2=NN(C=C2)C
This item is not returnable. View return policy
179873-47-3
C12H15N3
201.273
MFCD09702353
BBOCGPKUAXCMLG-UHFFFAOYSA-N
n-methyl-4-1-methyl-1h-pyrazol-3-yl benzylamine, benzenemethanamine,n-methyl-4-1-methyl-1h-pyrazol-3-yl, methyl 4-1-methylpyrazol-3-yl phenyl methyl amine, n-methyl-1-4-1-methyl-1h-pyrazol-3-yl phenyl methanamine, methyl 4-1-methyl-1h-pyrazol-3-yl phenyl methyl amine
24229474
N-methyl-1-[4-(1-methylpyrazol-3-yl)phenyl]methanamine
CNCC1=CC=C(C=C1)C2=NN(C=C2)C
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 201.27
Percent Purity 90%
Chemical Name or Material N-methyl-4-(1-methyl-1H-pyrazol-3-yl)benzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'