missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-(3-phenylisoxazol-5-yl)methylamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 188.23 |
Percent Purity | 97% |
Chemical Name or Material | N-methyl-(3-phenylisoxazol-5-yl)methylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'