missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-nitrobenzylamine, 95%, Thermo Scientific™

Catalog Number p-7055379
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
19499-61-7
C8H10N2O2
166.18
MFCD04115408
NTPAPKLGADEFAM-UHFFFAOYSA-N
n-methyl-1-3-nitrophenyl methanamine, n-methyl-n-3-nitrobenzyl amine, methyl 3-nitrophenyl methyl amine, benzenemethanamine, n-methyl-3-nitro, 3-nitro-n-methylbenzylamine, n-methyl-3-nitrobenzylamine, n-3-nitrophenylmethyl methylamine, n-methyl-1-3-nitrophenyl methylamine
409439
CNCC1=CC(=CC=C1)[N+](=O)[O-]
This item is not returnable. View return policy
19499-61-7
C8H10N2O2
166.18
MFCD04115408
NTPAPKLGADEFAM-UHFFFAOYSA-N
n-methyl-1-3-nitrophenyl methanamine, n-methyl-n-3-nitrobenzyl amine, methyl 3-nitrophenyl methyl amine, benzenemethanamine, n-methyl-3-nitro, 3-nitro-n-methylbenzylamine, n-methyl-3-nitrobenzylamine, n-3-nitrophenylmethyl methylamine, n-methyl-1-3-nitrophenyl methylamine
409439
CNCC1=CC(=CC=C1)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Methyl-3-nitrobenzylamine is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN2735
Solubility Information Slightly soluble in water.
Formula Weight 166.18
Percent Purity 95%
Physical Form Oil
Chemical Name or Material N-Methyl-3-nitrobenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'