missing translation for 'onlineSavingsMsg'
Learn More

n-methyl(3-methyl-2-furyl)methylamine, 90%, Maybridge

Brand:  Maybridge CC55546DA

Code : 31

Additional Details : CAS Number : 916766-86-4 Weight : 0.05100kg Transport : UN number : 2733 Chem class : 3 Pack group : III

Packaging Amber Glass Bottle
Quantity 1g

 View more versions of this product

Product Code. 10535744

  • £95.00 / 1g


Estimated Shipment date
from Supplier 27-07-2018
Add to basket

Specifications

Chemical Identifiers

N-methyl(3-methyl-2-furyl)methylamine
89.5
90%
C7H11NO
MFCD09879907
916766-86-4
100.0
90%
125.17

Specifications

N-methyl(3-methyl-2-furyl)methylamine
89.5
Amber Glass Bottle
90%
125.17
1g
916766-86-4
100.0
90%
C7H11NO
MFCD09879907
Videos
SDS