missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-[(6-methylpyrazin-2-yl)oxy]benzylamine, 97%, Maybridge

Brand:  Maybridge CC62646CB

Code : 31

Additional Details : CAS Number : 912569-66-5 Weight : 0.05025kg Transport : UN number : 2735 Chem class : 1b Pack group : III

Quantity 250mg
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10122754

  • £35.02 / 250mg


Estimated Shipment date
from Supplier 25-01-2018
Add to basket

Specifications

Chemical Identifiers

N-Methyl-3-[(6-methylpyrazin-2-yl)oxy]benzylamine
96.5
97%
C13H15N3O
MFCD09817528
912569-66-5
100.0
95%
229.28

Specifications

N-Methyl-3-[(6-methylpyrazin-2-yl)oxy]benzylamine
96.5
Amber Glass Bottle
95%
229.28
250mg
912569-66-5
100.0
97%
C13H15N3O
MFCD09817528
Videos
SDS