missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Methyl-3,5-dinitro-N-phenylbenzamide, 97%

Brand:  Alfa Aesar™ H59270.03

Code : NEW

Additional Details : CAS Number : 60965-27-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15445528

  • £358.49 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Methyl-3,5-dinitro-N-phenylbenzamide
301.25
2891007
60965-27-7
MFCD00791284

Specifications

N-Methyl-3,5-dinitro-N-phenylbenzamide
1g
Solid
301.25
2891007
Pale brown
60965-27-7
97%
MFCD00791284
Videos
SDS