missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-3-(1H-pyrazol-1-ylmethyl)benzylamine, 90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 201.27 |
Percent Purity | 90% |
Chemical Name or Material | N-methyl-3-(1H-pyrazol-1-ylmethyl)benzylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'