missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2-pyrrolidin-1-ylbenzylamine, 97%, Maybridge

Brand:  Maybridge CC44846EA

Code : 31

Additional Details : CAS Number : 871217-37-7 Weight : 0.06000kg Transport : UN number : 2922 Chem class : 1b Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 10663045

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

N-Methyl-2-pyrrolidin-1-ylbenzylamine
96.5
97%
C12H18N2
MFCD07348555
871217-37-7
100.0
97%
190.29

Specifications

N-Methyl-2-pyrrolidin-1-ylbenzylamine
871217-37-7
100.0
97%
190.29
10g
96.5
97%
C12H18N2
MFCD07348555
Videos
SDS