missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-2-morpholino-5-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
CAS | 886851-52-1 |
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Molecular Formula | C13H17F3N2O |
MDL Number | MFCD09025891 |
Quantity | 1 g |
Synonym | n-methyl-2-morpholino-5-trifluoromethyl benzylamine, methyl 2-morpholin-4-yl-5-trifluoromethyl phenyl methyl amine, n-methyl-n-2-morpholin-4-yl-5-trifluoromethyl benzyl amine, benzenemethanamine,n-methyl-2-4-morpholinyl-5-trifluoromethyl, n-methyl-1-2-morpholino-5-trifluoromethyl phenyl methanamine, n-methyl-1-2-morpholin-4-yl-5-trifluoromethyl phenyl methanamine |
InChI Key | UAZNBBVDDCKGES-UHFFFAOYSA-N |
SMILES | CNCC1=C(C=CC(=C1)C(F)(F)F)N1CCOCC1 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'