missing translation for 'onlineSavingsMsg'
Learn More

N-methyl-(2-methyl-1,3-oxazol-4-yl)methylamine, Thermo Scientific™

Catalog Number p-4391410
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
1065073-46-2
C6H10N2O
126.159
SFLPQWGHHRKLNJ-UHFFFAOYSA-N
methyl 2-methyl-1,3-oxazol-4-yl methyl amine, 4-methylaminomethyl-2-methyl-1,3-oxazole, 4-oxazolemethanamine, n,2-dimethyl, 2-methyl-4-methylamino methyl-1,3-oxazole, methyl 2-methyl 1,3-oxazol-4-yl methyl amine, n-methyl-1-2-methyl-1,3-oxazol-4-yl methanamine, n-methyl-1-2-methyl-1,3-oxazol-4-yl methylamine, n-methyl-2-methyl-1,3-oxazol-4-yl methylamine
44119607
N-methyl-1-(2-methyl-1,3-oxazol-4-yl)methanamine
CC1=NC(=CO1)CNC
This item is not returnable. View return policy
1065073-46-2
C6H10N2O
126.159
SFLPQWGHHRKLNJ-UHFFFAOYSA-N
methyl 2-methyl-1,3-oxazol-4-yl methyl amine, 4-methylaminomethyl-2-methyl-1,3-oxazole, 4-oxazolemethanamine, n,2-dimethyl, 2-methyl-4-methylamino methyl-1,3-oxazole, methyl 2-methyl 1,3-oxazol-4-yl methyl amine, n-methyl-1-2-methyl-1,3-oxazol-4-yl methanamine, n-methyl-1-2-methyl-1,3-oxazol-4-yl methylamine, n-methyl-2-methyl-1,3-oxazol-4-yl methylamine
44119607
N-methyl-1-(2-methyl-1,3-oxazol-4-yl)methanamine
CC1=NC(=CO1)CNC
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 126.16
Percent Purity 97%
Chemical Name or Material N-methyl-(2-methyl-1, 3-oxazol-4-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'