missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl[2-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methylamine, 95%, Thermo Scientific™

Catalog Number p-3541096
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
879896-56-7
C11H13N3O
203.25
MFCD09702360
LJDBGVKOXDSIQQ-UHFFFAOYSA-N
n-methyl 2-3-methyl-1,2,4-oxadiazol-5-yl phenyl methylamine, benzenemethanamine,n-methyl-2-3-methyl-1,2,4-oxadiazol-5-yl, methyl 2-3-methyl-1,2,4-oxadiazol-5-yl phenyl methyl amine, n-methyl-1-2-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanamine, methyl 2-3-methyl 1,2,4-oxadiazol-5-yl phenyl methyl amine
24229504
CNCC1=CC=CC=C1C1=NC(C)=NO1
This item is not returnable. View return policy
879896-56-7
C11H13N3O
203.25
MFCD09702360
LJDBGVKOXDSIQQ-UHFFFAOYSA-N
n-methyl 2-3-methyl-1,2,4-oxadiazol-5-yl phenyl methylamine, benzenemethanamine,n-methyl-2-3-methyl-1,2,4-oxadiazol-5-yl, methyl 2-3-methyl-1,2,4-oxadiazol-5-yl phenyl methyl amine, n-methyl-1-2-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanamine, methyl 2-3-methyl 1,2,4-oxadiazol-5-yl phenyl methyl amine
24229504
CNCC1=CC=CC=C1C1=NC(C)=NO1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name N-methyl-1-[2-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanamine
Formula Weight 203.24
Percent Purity 95%
Chemical Name or Material N-methyl[2-(3-methyl-1, 2, 4-oxadiazol-5-yl)phenyl]methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'