missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-(1-methyl-1h-indol-7-yl)methylamine, 90%, Maybridge

£74.00 - £202.00

Chemical Identifiers

CAS 709649-75-2
Molecular Formula C11H14N2
Molecular Weight (g/mol) 174.247
MDL Number MFCD11506353
InChI Key JERMRTDADLXHSD-UHFFFAOYSA-N
Synonym methyl 1-methylindol-7-yl methyl amine, n-methyl-1-1-methyl-1h-indol-7-yl methylamine, n-methyl-1-methyl-1h-indol-7-yl methylamine, 1h-indole-7-methanamine, n,1-dimethyl, n-methyl-1-1-methyl-1h-indol-7-yl methanamine, 1h-indole-7-methanamine,n,1-dimethyl, methyl-1-methyl-1h-indol-7-ylmethyl amine, 7-methylamino methyl-1-methyl-1h-indole, methyl 1-methyl-1h-indol-7-yl methyl amine
PubChem CID 33589520
IUPAC Name N-methyl-1-(1-methylindol-7-yl)methanamine
SMILES CNCC1=CC=CC2=C1N(C=C2)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10526154 SDS View Documents Maybridge
CC66946CB
250mg Amber Glass Bottle £74.00
250mg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-07-2020
Please to purchase this item. Need a web account? Register with us today!
10002184 SDS View Documents Maybridge
CC66946DA
1g Amber Glass Bottle £202.00
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-07-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

709649-75-2
174.247
JERMRTDADLXHSD-UHFFFAOYSA-N
33589520
CNCC1=CC=CC2=C1N(C=C2)C
C11H14N2
MFCD11506353
methyl 1-methylindol-7-yl methyl amine, n-methyl-1-1-methyl-1h-indol-7-yl methylamine, n-methyl-1-methyl-1h-indol-7-yl methylamine, 1h-indole-7-methanamine, n,1-dimethyl, n-methyl-1-1-methyl-1h-indol-7-yl methanamine, 1h-indole-7-methanamine,n,1-dimethyl, methyl-1-methyl-1h-indol-7-ylmethyl amine, 7-methylamino methyl-1-methyl-1h-indole, methyl 1-methyl-1h-indol-7-yl methyl amine
N-methyl-1-(1-methylindol-7-yl)methanamine

Specifications

N-methyl-(1-methyl-1H-indol-7-yl)methylamine
250mg
89.5
90%
MFCD11506353
JERMRTDADLXHSD-UHFFFAOYSA-N
N-methyl-1-(1-methylindol-7-yl)methanamine
33589520
90%
Amber Glass Bottle
709649-75-2
100.0
C11H14N2
methyl 1-methylindol-7-yl methyl amine, n-methyl-1-1-methyl-1h-indol-7-yl methylamine, n-methyl-1-methyl-1h-indol-7-yl methylamine, 1h-indole-7-methanamine, n,1-dimethyl, n-methyl-1-1-methyl-1h-indol-7-yl methanamine, 1h-indole-7-methanamine,n,1-dimethyl, methyl-1-methyl-1h-indol-7-ylmethyl amine, 7-methylamino methyl-1-methyl-1h-indole, methyl 1-methyl-1h-indol-7-yl methyl amine
CNCC1=CC=CC2=C1N(C=C2)C
174.247
174.25