missing translation for 'onlineSavingsMsg'
Learn More

N-Methoxy-N-methylbenzamide, 98%, ACROS Organics™

Brand:  Acros Organics 436560250

Code : 29

Additional Details : CAS Number : 6919-61-5 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 11945551

  • £192.60 / 25g


Estimated Shipment
15-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Methoxy-N-methylbenzamide
97.5
≥97.5%
C9H11NO2
6919-61-5
100.0
98%
165.19

Specifications

N-Methoxy-N-methylbenzamide
25g
97.5
Authentic
Liquid
98%
165.19
Colorless to yellow
6919-61-5
100.0
Glass bottle
≥97.5%
C9H11NO2
Videos
SDS