missing translation for 'onlineSavingsMsg'
Learn More

N-Methoxy-N-methylacetamide, 98%, Thermo Scientific Chemicals

Catalog Number p-2691943
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
78191-00-1
C4H9NO2
103.12
MFCD00060098
OYVXVLSZQHSNDK-UHFFFAOYSA-N
n-methyl-n-methoxyacetamide, acetamide, n-methoxy-n-methyl, n-methoxyl-n-methylacetamide, n-methoxy-n-methyl-acetamide, n-methoxy-n-methylacetamid, pubchem12775, acmc-209pdo, n-methoxy-n-methylacetamde, n-methoxy n-methylacetamide, n-methoxy-n-methyl acetamide
537505
N-methoxy-N-methylacetamide
CON(C)C(C)=O
This item is not returnable. View return policy
78191-00-1
C4H9NO2
103.12
MFCD00060098
OYVXVLSZQHSNDK-UHFFFAOYSA-N
n-methyl-n-methoxyacetamide, acetamide, n-methoxy-n-methyl, n-methoxyl-n-methylacetamide, n-methoxy-n-methyl-acetamide, n-methoxy-n-methylacetamid, pubchem12775, acmc-209pdo, n-methoxy-n-methylacetamde, n-methoxy n-methylacetamide, n-methoxy-n-methyl acetamide
537505
N-methoxy-N-methylacetamide
CON(C)C(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9700g/mL
Boiling Point 152°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CON(OCH3)CH3
Refractive Index 1.4240 to 1.426
Quantity 5 g
Specific Gravity 0.97
Solubility Information Solubility in water: soluble
Formula Weight 103.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Methoxy-N-methylacetamide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • N-Methoxy-N-methylacetamide
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'