missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Lauroylsarcosine sodium salt, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 94 |
CAS Max % | 100 |
Melting Point | 45.0°C to 47.0°C |
Color | White |
pH | 7.0 to 9.0 |
Loss on Drying | 7% max. () |
Infrared Spectrum | Authentic |
Assay Percent Range | 0.95 |
Packaging | Plastic Drum |
Linear Formula | NaCO2CH2N(CH3)CO(CH2)10CH3 |
Show More |
Safety and Handling
Product Identifier
- N-Lauroylsarcosine sodium salt
- Danger
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H330-Fatal if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'