missing translation for 'onlineSavingsMsg'
Learn More

N-Lauroylsarcosine sodium salt, 95%, Thermo Scientific Chemicals

Catalog Number p-7041727
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
137-16-6
C15H28NNaO3
293.38
MFCD00042728
KSAVQLQVUXSOCR-UHFFFAOYSA-M
sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
23668817
sodium;2-[dodecanoyl(methyl)amino]acetate
[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O
This item is not returnable. View return policy
137-16-6
C15H28NNaO3
293.38
MFCD00042728
KSAVQLQVUXSOCR-UHFFFAOYSA-M
sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
23668817
sodium;2-[dodecanoyl(methyl)amino]acetate
[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

N-Lauroylsarcosine is used for solubilization and separation of membrane proteins and glycoprotein's; reported to inhibit hexokinase. It is useful in concentrated salt solutions used in the cell lysis step during RNA purification (helps avoid excessive foaming). It has been used to indicate paramagnetic anisotropy sign change in micelle mesophage. Inhibits bacterial flora of human saliva/gut at 0.25% as well as acting as a fungi static agent in aqueous dispersion (1%).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Lauroylsarcosine is used for solubilization and separation of membrane proteins and glycoprotein′s; reported to inhibit hexokinase. It is useful in concentrated salt solutions used in the cell lysis step during RNA purification (helps avoid excessive foaming). It has been used to indicate paramagnetic anisotropy sign change in micelle mesophage. Inhibits bacterial flora of human saliva/gut at 0.25% as well as acting as a fungi static agent in aqueous dispersion (1%).

Solubility
Soluble in water (293 g/L).

Notes
Hygroscopic, store away form water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent and reducing agent.
TRUSTED_SUSTAINABILITY
Melting Point 44°C to 47°C
Color White
Linear Formula CH3(CH2)10CON(CH3)CH2COONa
Quantity 10 g
Beilstein 5322974
Sensitivity Hygroscopic
Merck Index 14,4368
Solubility Information Soluble in water (293g/L).
Formula Weight 293.38
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material N-Lauroylsarcosine sodium salt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'